missing translation for 'onlineSavingsMsg'
Learn More

1,2,2,6,6-Pentamethylpiperidine, 97%

Product Code. 11392859
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11392859 1 g 1g
11302869 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11392859 Supplier Thermo Scientific Alfa Aesar Supplier No. H27729.03
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 79-55-0 | C10H21N | 155.285 g/mol

Pharmacological evaluation of the antagonism of nicotine's central effects by mecamylamine and pempidine where pempidine was found to increase the ED50 of nicotine (0.73 mg/kg) for depression of spontaneous activity in a dose-related manner. At a dose of 3 mg/kg, pempidine increased nicotine's ED50 4.7-fold.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Pharmacological evaluation of the antagonism of nicotine′s central effects by mecamylamine and pempidine where pempidine was found to increase the ED50 of nicotine (0.73 mg/kg) for depression of spontaneous activity in a dose-related manner. At a dose of 3 mg/kg, pempidine increased nicotine′s ED50 4.7-fold.

Solubility
Soluble in toluene, dimethylformamide.

Notes
Air Sensitive. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents, air.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 79-55-0
Molecular Formula C10H21N
Molecular Weight (g/mol) 155.285
MDL Number MFCD00006493
InChI Key XULIXFLCVXWHRF-UHFFFAOYSA-N
Synonym pempidine, pyrilene, perolysen, tenormal, piperidine, 1,2,2,6,6-pentamethyl, pempidina dcit, pempidine inn:ban, pempidinum inn-latin, unii-n5i18ji9d6, 1,2,2,6,6-pentamethyl-piperidine
PubChem CID 6603
IUPAC Name 1,2,2,6,6-pentamethylpiperidine
SMILES CC1(CCCC(N1C)(C)C)C

Specifications

Density 0.858
Boiling Point 187°C to 188°C
Flash Point 50°C (122°F)
Odor Amine-like
Quantity 1 g
UN Number UN1992
Beilstein 103806
Sensitivity Air Sensitive
Merck Index 14,7080
Solubility Information Soluble in toluene,dimethylformamide.
Formula Weight 155.28
Percent Purity 97%
Chemical Name or Material 1,2,2,6,6-Pentamethylpiperidine
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • 1,2,2,6,6-Pentamethylpiperidine
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 3
  • Serious eye damage/eye irritation Category 2
  • Flammable liquid Category 3
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H226-Flammable liquid and vapour.
  • H301-Toxic if swallowed.
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
Supplemental information
  • MIXTURE LIST-Contain: 1,2,2,6,6-Pentamethylpiperidine

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.