missing translation for 'onlineSavingsMsg'
Learn More

1-(2,2-Difluorobenzo[1,3]dioxol-5-yl)cyclopropanecarboxylic Acid, TRC

Product Code. 30383429
Change view
Click to view available options
Quantity:
5 g
Unit Size:
5g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30383429 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30383429 Supplier Toronto Research Chemicals Supplier No. TRCD4456735G
 Restricted Item
Estimated Shipment: 17-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 1-(2,2-Difluorobenzo[1,3]dioxol-5-yl)cyclopropanecarboxylic Acid
CAS 862574-88-7
Molecular Formula C11H8O4F2
Quantity 5 g
Synonym 1-(2,2-Difluoro-1,3-benzodioxol-5-yl)cyclopropanecarboxylic Acid, 1-(2,2-Difluoro-1,3-benzodioxol-5-yl)cyclopropane-1-carboxylic Acid, 1-(2,2-Difluoro-2H-1,3-benzodioxol-5-yl)cyclopropane-1-carboxylic Acid, 1-(2,2-Difluorobenzo[d][1,3]dioxol-5-yl)cyclopropanecarboxylic Acid, 1-(2,2-Difluorobenzodioxol-5-yl)cyclopropanecarboxylic Acid
InChI Formula InChI=1S/C11H8F2O4/c12-11(13)16-7-2-1-6(5-8(7)17-11)10(3-4-10)9(14)15/h1-2,5H,3-4H2,(H,14,15)
SMILES OC(=O)C1(CC1)c2ccc3OC(F)(F)Oc3c2
IUPAC Name 1-(2,2-difluoro-1,3-benzodioxol-5-yl)cyclopropane-1-carboxylic acid
Molecular Weight (g/mol) 242.17
Formula Weight 242.0391
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.