missing translation for 'onlineSavingsMsg'
Learn More

1,2,3,4,5-Pentachlorobenzene-13C6, TRC

Product Code. 30012331
Change view
Click to view available options
Quantity:
1 mg
10 mg
Unit Size:
10mg
1mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30012331 10 mg 10mg
30004470 1 mg 1mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30012331 Supplier Toronto Research Chemicals Supplier No. TRCP23792210MG
 Restricted Item
Estimated Shipment: 14-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2483735-54-0
Molecular Formula 13C6 H Cl5
Molecular Weight (g/mol) 256.29
Synonym Pentachlorobenzene 13C6, Pentachlorobenzene-13C6, [13C6]Pentachlorobenzene, 1,2,3,4,5-Pentachlorobenzene-1,2,3,4,5,6-13C6
IUPAC Name 1,2,3,4,5-pentachloro(1,2,3,4,5,6-13C6)cyclohexa-1,3,5-triene
SMILES Cl[13c]1[13cH][13c](Cl)[13c](Cl)[13c](Cl)[13c]1Cl

Specifications

Quantity 10 mg
InChI Formula InChI=1S/C6HCl5/c7-2-1-3(8)5(10)6(11)4(2)9/h1H/i1+1,2+1,3+1,4+1,5+1,6+1
Formula Weight 253.872
Percent Purity >95%
Chemical Name or Material Pentachlorobenzene 13C6
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.