missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals 1,2,3,4,6-Penta-O-acetyl-alpha-D-mannopyranose, 98%

Product Code. 15408139
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
This item is not returnable. View return policy

Product Code. 15408139

Brand: Thermo Scientific Chemicals J66685.06

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 4163-65-9 | C16H22O11 | 390.341 g/mol

α-D-Mannose pentaacetate is used for glycosylation in a study that assessed novel synthetic inhibitors of selectin-mediated cell adhesion. It has also been used in a study to investigate stereospecific entry to spiroketal glycosides using alkylidenecarbene C-H insertion.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
α-D-Mannose pentaacetate is used for glycosylation in a study that assessed novel synthetic inhibitors of selectin-mediated cell adhesion. It has also been used in a study to investigate stereospecific entry to spiroketal glycosides using alkylidenecarbene C-H insertion.

Solubility
Soluble in chloroform

Notes
Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 4163-65-9
Molecular Formula C16H22O11
Molecular Weight (g/mol) 390.341
MDL Number MFCD00069798
InChI Key LPTITAGPBXDDGR-OWYFMNJBSA-N
Synonym a-d-mannose pentaacetate, alpha-d-mannose pentaacetate, 1,2,3,4,6-penta-o-acetyl-alpha-d-mannopyranose, alpha-d-mannopyranose pentaacetate, 1-o,2-o,3-o,4-o,6-o-pentaacetyl-a-d-mannopyranose, man oac 5, a-d-mannosepentaacetate, pubchem22277, 1,2,3,4,6-penta-o-acetyl-, a-d-mannopyranose, 1,2,3,4,6-penta-o-acetyl-a-d-mannopyranose
PubChem CID 11741089
IUPAC Name [(2R,3R,4S,5S,6R)-3,4,5,6-tetraacetyloxyoxan-2-yl]methyl acetate
SMILES CC(=O)OCC1C(C(C(C(O1)OC(=O)C)OC(=O)C)OC(=O)C)OC(=O)C

Specifications

Color White
Quantity 5 g
Solubility Information Soluble in chloroform
Optical Rotation +54° (c=1 in Chloroform)
Formula Weight 390.34
Percent Purity 98%
Physical Form Powder
Chemical Name or Material 1,2,3,4,6-Penta-O-acetyl-alpha-D-mannopyranose

RUO – Research Use Only

Corrección del contenido de un producto

Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.

Título del producto

Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.

Gracias por ayudarnos a mejorar nuestra web. Su comentario ha sido enviado