missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals 1,2,3,4,6-Penta-O-acetyl-alpha-D-mannopyranose, 98%

Product Code. 15418139
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15418139 25 g 25g
15408139 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15418139 Supplier Thermo Scientific Chemicals Supplier No. J66685.14
 Restricted Item
Estimated Shipment: 30-12-2026
Lägg i varukorgen
Lägg i varukorgen
Request Bulk or Custom Format

missing translation for 'validMainframeMsg'

Denna artikel kan inte returneras. Se returpolicy

CAS: 4163-65-9 | C16H22O11 | 390.341 g/mol

α-D-Mannose pentaacetate is used for glycosylation in a study that assessed novel synthetic inhibitors of selectin-mediated cell adhesion. It has also been used in a study to investigate stereospecific entry to spiroketal glycosides using alkylidenecarbene C-H insertion.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
α-D-Mannose pentaacetate is used for glycosylation in a study that assessed novel synthetic inhibitors of selectin-mediated cell adhesion. It has also been used in a study to investigate stereospecific entry to spiroketal glycosides using alkylidenecarbene C-H insertion.

Solubility
Soluble in chloroform

Notes
Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
TRUSTED_SUSTAINABILITY

Kemiska identifierare

CAS 4163-65-9
Molecular Formula C16H22O11
Molecular Weight (g/mol) 390.341
MDL Number MFCD00069798
InChI Key LPTITAGPBXDDGR-OWYFMNJBSA-N
Synonym a-d-mannose pentaacetate, alpha-d-mannose pentaacetate, 1,2,3,4,6-penta-o-acetyl-alpha-d-mannopyranose, alpha-d-mannopyranose pentaacetate, 1-o,2-o,3-o,4-o,6-o-pentaacetyl-a-d-mannopyranose, man oac 5, a-d-mannosepentaacetate, pubchem22277, 1,2,3,4,6-penta-o-acetyl-, a-d-mannopyranose, 1,2,3,4,6-penta-o-acetyl-a-d-mannopyranose
PubChem CID 11741089
IUPAC Name [(2R,3R,4S,5S,6R)-3,4,5,6-tetraacetyloxyoxan-2-yl]methyl acetate
SMILES CC(=O)OCC1C(C(C(C(O1)OC(=O)C)OC(=O)C)OC(=O)C)OC(=O)C

Specifikationer

Color White
Quantity 25 g
Solubility Information Soluble in chloroform
Optical Rotation +54° (c=1 in Chloroform)
Formula Weight 390.34
Percent Purity 98%
Physical Form Powder
Chemical Name or Material 1,2,3,4,6-Penta-O-acetyl-alpha-D-mannopyranose

RUO – Research Use Only

Produkttitel
Välj ett problem

Genom att klicka på Skicka bekräftar du att du kan bli kontaktad av Fisher Scientific angående feedbacken du har lämnat i detta formulär. Vi kommer inte att dela din information för andra ändamål. All kontaktinformation som tillhandahålls ska också underhållas i enlighet med vår Sekretesspolicy.