missing translation for 'onlineSavingsMsg'
Learn More

1,2,3,4-Tetra-O-acetyl-Beta-D-glucopyranose, TRC

Product Code. 30135691
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30135691 5 g 5g
30133166 25 g 25g
30011464 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30135691 Supplier Toronto Research Chemicals Supplier No. TRCT2800055G
 Restricted Item
Estimated Shipment: 14-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 13100-46-4
Molecular Formula C14 H20 O10
Molecular Weight (g/mol) 348.3
Synonym β-D-Glucopyranose, 1,2,3,4-tetraacetate, Glucopyranose, 1,2,3,4-tetraacetate, β-D- (8CI), 1,2,3,4-Tetra-O-acetyl-β-D-glucopyranose, 1,2,3,4-Tetra-O-acetyl-β-D-glucose, NSC 409250
IUPAC Name [(2R,3R,4S,5R,6S)-4,5,6-triacetyloxy-2-(hydroxymethyl)oxan-3-yl] acetate
SMILES CC(=O)O[C@@H]1O[C@H](CO)[C@@H](OC(=O)C)[C@H](OC(=O)C)[C@H]1OC(=O)C

Specifications

Quantity 5 g
InChI Formula InChI=1S/C14H20O10/c1-6(16)20-11-10(5-15)24-14(23-9(4)19)13(22-8(3)18)12(11)21-7(2)17/h10-15H,5H2,1-4H3/t10-,11-,12+,13-,14-/m1/s1
Formula Weight 348.1056
Chemical Name or Material 1,2,3,4-Tetra-O-acetyl-beta-D-glucopyranose
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.