missing translation for 'onlineSavingsMsg'
Learn More

1,2,3,4-Tetrahydro-1-naphthoic Acid, TRC

Product Code. 30134652
Change view
Click to view available options
Quantity:
1 g
10 g
Unit Size:
10g
1g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30134652 10 g 10g
30009332 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30134652 Supplier Toronto Research Chemicals Supplier No. TRCT29410510G
 Restricted Item
Estimated Shipment: 30-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1914-65-4
Molecular Formula C11 H12 O2
Molecular Weight (g/mol) 176.21
Synonym 1-Naphthalenecarboxylic acid, 1,2,3,4-tetrahydro-, 1-Naphthoic acid, 1,2,3,4-tetrahydro- (6CI,7CI,8CI), 1,2,3,4-Tetrahydro-1-naphthalenecarboxylic acid, (±)-1,2,3,4-Tetrahydronaphthalene-1-carboxylic acid, 1,2,3,4-Tetrahydro-1-naphthoic acid, 1,2,3,4-Tetrahydro-α-naphthoic acid, 1,2,3,4-Tetrahydronaphthalene-1-carboxylic acid, 1-Tetralincarboxylic acid
IUPAC Name 1,2,3,4-tetrahydronaphthalene-1-carboxylic acid
SMILES OC(=O)C1CCCc2ccccc12

Specifications

Quantity 10 g
InChI Formula InChI=1S/C11H12O2/c12-11(13)10-7-3-5-8-4-1-2-6-9(8)10/h1-2,4,6,10H,3,5,7H2,(H,12,13)
Formula Weight 176.0837
Chemical Name or Material 1,2,3,4-Tetrahydro-1-naphthalenecarboxylic Acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.