missing translation for 'onlineSavingsMsg'
Learn More

1,2,3,4-Tetrahydro-6,7-isoquinolinediol hydrobromide, 98%, Thermo Scientific™

Product Code. 10761891 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
Packaging:
Glass bottle
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10761891 1 g Glass bottle 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10761891 Supplier Thermo Scientific Acros Supplier No. 235060010
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

Specifications

CAS 52768-23-7
CAS Min % 97.5
CAS Max % 100.0
Melting Point 210°C
Color Beige to Brown
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (Argentometry)
Packaging Glass bottle
Molecular Formula C9H12NO2
MDL Number MFCD00078115
Quantity 1 g
Synonym 1,2,3,4-tetrahydroisoquinoline-6,7-diol hydrobromide, 1,2,3,4-tetrahydro-6,7-isoquinolinediol hydrobromide, 1,2,3,4-tetrahydro isoquinoline-6,7-diol hydrobromide, norsalsolinol hydrobromide, 1,2,3,4-tetrahydroisoquinoline-6,7-diol, bromide, 1,2,3,4-tetrahydroisoquinoline-6,7-diol hcl, 6,7-dihydroxytetrahydroisoquinoline hydrobromide, 6,7-dihydroxy-1,2,3,4-tetrahydroisoquinoline hydrobromide, 6,7-dihydroxy-1,2,3,4-tetrahydroisoquinolinehydrobromide, 6,7-isoquinolinediol, 1,2,3,4-tetrahydro-, hydrobromide
InChI Key MBFUSGLXKQWVDW-UHFFFAOYSA-O
SMILES OC1=CC2=C(C[NH2+]CC2)C=C1O
IUPAC Name 1,2,3,4-tetrahydroisoquinoline-6,7-diol;hydrobromide
Molecular Weight (g/mol) 166.20
PubChem CID 2724460
Formula Weight 246.1
Percent Purity 98%
Physical Form Powder
Chemical Name or Material 1,2,3,4-Tetrahydro-6,7-isoquinolinediol hydrobromide, 98%
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.