missing translation for 'onlineSavingsMsg'
Learn More

1,2,3,4-Tetrahydro-6-quinolinecarboxylic acid, 97%, Thermo Scientific™

Product Code. 10181022
Change view
Click to view available options
Quantity:
1 g
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
10181022 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10181022 Supplier Thermo Scientific Maybridge Supplier No. MO00019DA
 Restricted Item
Estimated Shipment: 12-01-2027
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Specifications

CAS 5382-49-0
CAS Min % 97.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Molecular Formula C10H11NO2
MDL Number MFCD00957087
Quantity 1 g
Synonym 1,2,3,4-tetrahydro-6-quinolinecarboxylic acid, 1,2,3,4-tetrahydro-quinoline-6-carboxylic acid, 6-quinolinecarboxylic acid, 1,2,3,4-tetrahydro, maybridge3_004537, 1,2,3,4-tetrahydro-6-quinolinecarboxylicacid, 1,2,3,4-tetrahydroquinolin-6-carboxylic acid
InChI Key ARNALYPZOYPNAF-UHFFFAOYSA-N
SMILES OC(=O)C1=CC2=C(NCCC2)C=C1
IUPAC Name 1,2,3,4-tetrahydroquinoline-6-carboxylic acid
Molecular Weight (g/mol) 177.20
PubChem CID 2779641
Formula Weight 177.2
Percent Purity 97%
Chemical Name or Material 1,2,3,4-Tetrahydro-6-quinolinecarboxylic acid
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.