missing translation for 'onlineSavingsMsg'
Learn More

1,2,3,4-Tetrahydroisoquinoline-6,7-diol Hydrobromide, TRC

Product Code. 30456983
Change view
Click to view available options
Quantity:
10 mg
50 mg
100 mg
Unit Size:
10mg
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30456983 10 mg 10mg
30454740 50 mg 50mg
30455002 100 mg -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30456983 Supplier Toronto Research Chemicals Supplier No. TRCT79748810MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 52768-23-7
Molecular Formula C9H11NO2.HBr
Molecular Weight (g/mol) 246.1
IUPAC Name 1,2,3,4-tetrahydroisoquinoline-6,7-diol;hydrobromide
SMILES Br.Oc1cc2CCNCc2cc1O

Specifications

Quantity 10 mg
InChI Formula InChI=1S/C9H11NO2.BrH/c11-8-3-6-1-2-10-5-7(6)4-9(8)12;/h3-4,10-12H,1-2,5H2;1H
Formula Weight 245.0051
Chemical Name or Material 1,2,3,4-Tetrahydroisoquinoline-6,7-diol Hydrobromide
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.