missing translation for 'onlineSavingsMsg'
Learn More

1,2,3,4-Tetrahydroisoquinoline, 95%

Product Code. 10520224 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
Packaging:
Glass bottle
Unit Size:
100g
25g
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Code produit Quantity Packaging unitSize
10520224 500 g Glass bottle 500g
10635572 25 g Glass bottle 25g
10295420 100 g Glass bottle 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Les retours ne sont pas autorisés pour ce produit. Afficher la politique du retour.
Code produit 10520224 Fournisseur Thermo Scientific Acros Code fournisseur 138205000
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
Voir les produits de remplacement

Les retours ne sont pas autorisés pour ce produit. Afficher la politique du retour.

CAS: 91-21-4 | C9H11N | 133.19 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Identifiants chimiques

CAS 91-21-4
Molecular Formula C9H11N
Molecular Weight (g/mol) 133.19
MDL Number MFCD00006896
InChI Key UWYZHKAOTLEWKK-UHFFFAOYSA-N
Synonym tetrahydroisoquinoline, 1,2,3,4-tetrahydro-isoquinoline, isoquinoline, 1,2,3,4-tetrahydro, 1,2,3,4-tetrahydro isoquinoline, 1,2,3,4-tetrahydro-2-isoquinoline, 1,2,3,4-tetrahydro-2-azanaphthalene, unii-56w89fbx3e, chembl14346, 3,4-dihydro-1h-isoquinoline, tetrahydroisoquinolin
PubChem CID 7046
IUPAC Name 1,2,3,4-tetrahydroisoquinoline
SMILES C1CC2=CC=CC=C2CN1

Spécification

CAS Min % 94.0
CAS Max % 100.0
Melting Point -30°C
Color Yellow to Brown
Density 1.0600g/mL
Boiling Point 232°C to 233°C
Flash Point 98°C
Infrared Spectrum Authentic
Assay Percent Range 94% min. (GC)
Packaging Glass bottle
Refractive Index 1.5665 to 1.5685
Quantity 500 g
Beilstein 20, 275
Specific Gravity 1.06
Solubility Information Solubility in water: 20g/L (20°C). Other solubilities: soluble in ethyl alcohol,benzene,acids
Formula Weight 133.19
Percent Purity 95%
Physical Form Liquid
Chemical Name or Material 1,2,3,4-Tetrahydroisoquinoline
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • 1,2,3,4-Tetrahydroisoquinoline
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
  • H312-Harmful in contact with skin.
  • H332-Harmful if inhaled.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.