missing translation for 'onlineSavingsMsg'
Learn More

1,2,3,4-Tetrahydroisoquinoline, 97%

Product Code. 11348005
Change view
Click to view available options
Quantity:
25 g
100 g
Unit Size:
100g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11348005 100 g 100g
11338005 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11348005 Supplier Thermo Scientific Alfa Aesar Supplier No. L08143.22
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 91-21-4 | C9H11N | 133.19 g/mol

It is used as a reagent in the preparation of4-(1,2,4-oxadiazol-5-yl)piperidine-1-carboxamides as antiproliferative tubulin inhibitors.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is used as a reagent in the preparation of4-(1,2,4-oxadiazol-5-yl)piperidine-1-carboxamides as antiproliferative tubulin inhibitors.

Solubility
Soluble in water at 20°C 20g/L.

Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition. Protect from heat. Keep it refrigerated.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 91-21-4
Molecular Formula C9H11N
Molecular Weight (g/mol) 133.19
MDL Number MFCD00006896
InChI Key UWYZHKAOTLEWKK-UHFFFAOYSA-N
Synonym tetrahydroisoquinoline, 1,2,3,4-tetrahydro-isoquinoline, isoquinoline, 1,2,3,4-tetrahydro, 1,2,3,4-tetrahydro isoquinoline, 1,2,3,4-tetrahydro-2-isoquinoline, 1,2,3,4-tetrahydro-2-azanaphthalene, unii-56w89fbx3e, chembl14346, 3,4-dihydro-1h-isoquinoline, tetrahydroisoquinolin
PubChem CID 7046
IUPAC Name 1,2,3,4-tetrahydroisoquinoline
SMILES C1CC2=CC=CC=C2CN1

Specifications

Melting Point -30°C
Density 1.049
pH 11
Boiling Point 232°C to 233°C
Flash Point 98°C (208°F)
Odor Amine-like
Refractive Index 1.568
Quantity 100 g
Beilstein 116156
Solubility Information Soluble in water at 20°C 20g/L.
Formula Weight 133.19
Percent Purity 97%
Chemical Name or Material 1,2,3,4-Tetrahydroisoquinoline
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.