missing translation for 'onlineSavingsMsg'
Learn More

1,2,3,4-Tetrahydroquinoline, 98%

Product Code. 11469767
Click to view available options
Quantity:
100 g
500 g
Unit Size:
100g
500g
This item is not returnable. View return policy

Product Code. 11469767

Brand: Thermo Scientific Alfa Aesar B22750.36

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 635-46-1 | C9H11N | 133.19 g/mol

1,2,3,4-Tetrahydroquinoline is used as an antioxidant and corrosion inhibitor. It is an active component in various dyes.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1,2,3,4-Tetrahydroquinoline is used as an antioxidant and corrosion inhibitor. It is an active component in various dyes.

Solubility
Miscible with alcohol, ether, carbon disulfide and many organic solvents. Slightly miscible with water.

Notes
Hygroscopic. Incomaptible with strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 635-46-1
Molecular Formula C9H11N
Molecular Weight (g/mol) 133.19
MDL Number MFCD00006693
InChI Key LBUJPTNKIBCYBY-UHFFFAOYSA-N
Synonym tetrahydroquinoline, 1,2,3,4-tetrahydro-quinoline, kusol, quinoline, 1,2,3,4-tetrahydro, unii-ccr50n1z9g, 1,2,3,4-tetrahydrochinoline, 1,2,3,4-tetrahydro quinoline, benzopiperidine, ccr50n1z9g, 1,2,3,4,-tetrahydroquinoline
PubChem CID 69460
ChEBI CHEBI:213323
IUPAC Name 1,2,3,4-tetrahydroquinoline
SMILES C1CNC2=CC=CC=C2C1

Specifications

Melting Point ∼14°C
Density 1.059
Boiling Point 248°C to 250°C
Flash Point 100°C (212°F)
Refractive Index 1.595
Quantity 500 g
Beilstein 116149
Solubility Information Miscible with alcohol,ether,carbon disulfide and many organic solvents. Slightly miscible with water.
Formula Weight 133.19
Percent Purity 99%
Chemical Name or Material 1,2,3,4-Tetrahydroquinoline
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.