missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1,2,3,4-Tetramethyl-1,3-cyclopentadiene, 85%, mixture of isomers, Thermo Scientific Chemicals
Shop All Thermo Scientific Chemicals ProductsBeskrivelse
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Tekniske data
Tekniske data
| Color | Yellow |
| Density | 0.8080g/mL |
| Boiling Point | 142.0°C |
| Flash Point | 42°C |
| Infrared Spectrum | Authentic |
| Assay Percent Range | 80% min. (GC) |
| Packaging | Glass bottle |
| Molecular Formula | C9H14 |
| Refractive Index | 1.4700 to 1.4760 |
| Quantity | 25 g |
| Vis mere |
Sikkerhed og håndtering
Produktidentifikator
- 1,2,3,4-Tetramethyl-1,3-cyclopentadiene
- Warning
- Flammable liquid Category 3
- H226-Flammable liquid and vapour.
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
RUO – Research Use Only
Produkttitel
Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.
Ser du en mulighed for forbedring?