missing translation for 'onlineSavingsMsg'
Learn More

1,2,3-Hexanetriol, 97%, Thermo Scientific™

Product Code. 15790839 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
Unit Size:
5g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15790839 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15790839 Supplier Thermo Scientific Acros Supplier No. 232510050
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
Se alternative produkter

Denne vare kan ikke returneres. Se returpolicy

Kemiske identifikatorer

CAS 90325-47-6
Molecular Formula C6H14O3
Molecular Weight (g/mol) 134.18
InChI Key XYXCXCJKZRDVPU-UHFFFAOYSA-N
Synonym 1,2,3-hexanetriol, rac-syn-hexane-1,2,3-triol, dsstox_cid_31384, dsstox_gsid_52884, 1,2,3-hexanetriol solution, 1,2,3-hexanetriol 1 m solution, 1,2,3-hexanetriol, bioxtra gc, additive screening solution 07/fluka kit no 78374
PubChem CID 2724429
IUPAC Name hexane-1,2,3-triol
SMILES CCCC(C(CO)O)O

Tekniske data

Melting Point 67.0°C to 69.0°C
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Linear Formula CH3(CH2)2CHOHCHOHCH2OH
Quantity 5 g
Solubility Information Solubility in water: miscible in water.
Formula Weight 134.18
Percent Purity 97%
Chemical Name or Material 1,2,3-Hexanetriol

RUO – Research Use Only

Produkttitel
Vælg et problem

Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.