missing translation for 'onlineSavingsMsg'
Learn More

1,2,3-Hexanetriol, 97%

Product Code. 11496330
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
Product Code. Quantity unitSize
11496330 5 g 5g
11486330 1 g 1g
2 options
This item is not returnable. View return policy

Product Code. 11496330

Brand: Thermo Scientific Alfa Aesar H27180.06

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 90325-47-6 | C6H14O3 | 134.175 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 90325-47-6
Molecular Formula C6H14O3
Molecular Weight (g/mol) 134.175
MDL Number MFCD00043071
InChI Key XYXCXCJKZRDVPU-UHFFFAOYSA-N
Synonym 1,2,3-hexanetriol, rac-syn-hexane-1,2,3-triol, dsstox_cid_31384, dsstox_gsid_52884, 1,2,3-hexanetriol solution, 1,2,3-hexanetriol 1 m solution, 1,2,3-hexanetriol, bioxtra gc, additive screening solution 07/fluka kit no 78374
PubChem CID 2724429
IUPAC Name hexane-1,2,3-triol
SMILES CCCC(C(CO)O)O

Specifications

Melting Point 64°C to 67°C
Quantity 5 g
Sensitivity Hygroscopic
Formula Weight 134.18
Percent Purity 97%
Chemical Name or Material 1,2,3-Hexanetriol

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.