missing translation for 'onlineSavingsMsg'
Learn More

1,2,3-Thiadiazole-4-carbaldehyde, Thermo Scientific™

Product Code. 10505954
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
Packaging:
Amber Glass Bottle
Unit Size:
1g
250mg
5g
Product Code. Quantity Packaging unitSize
10505954 5 g Amber Glass Bottle 5g
10019531 250 mg Amber Glass Bottle 250mg
10335012 1 g Amber Glass Bottle 1g
3 options
This item is not returnable. View return policy

Product Code. 10505954

Brand: Thermo Scientific Maybridge SP01373DE

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 27643-15-8
Molecular Formula C3H2N2OS
Molecular Weight (g/mol) 114.122
InChI Key JNEBZFFTOLBIKJ-UHFFFAOYSA-N
Synonym 1,2,3-thiadiazole-4-carbaldehyde, 1,2,3-thiadiazole-4-carboxaldehyde, 4-thiadiazolecarboxaldehyde, acmc-1cb1x, 1,2,3-thiadiazol-4-carboxaldehyde
PubChem CID 2737271
IUPAC Name thiadiazole-4-carbaldehyde
SMILES C1=C(N=NS1)C=O

Specifications

Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 5 g
Formula Weight 114.13
Percent Purity 97%
Chemical Name or Material 1, 2, 3-Thiadiazole-4-carbaldehyde
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.