missing translation for 'onlineSavingsMsg'
Learn More

1,2,3-Thiadiazole-4-carboxylic acid, 97%

Product Code. 11465786
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11465786 1 g 1g
11475786 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11465786 Supplier Thermo Scientific Alfa Aesar Supplier No. B20842.03
 Restricted Item
Estimated Shipment: 13-08-2026
Add to basket
This item is not returnable. View return policy

CAS: 4100-13-4 | C3H2N2O2S | 130.121 g/mol

It is used as an active pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is used as an active pharmaceutical intermediate.

Solubility
Slightly soluble in water.

Notes
Store away from oxidizing agents, reducing agents and bases. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 4100-13-4
Molecular Formula C3H2N2O2S
Molecular Weight (g/mol) 130.121
MDL Number MFCD00052099
InChI Key HJZYBDPHAHGHAZ-UHFFFAOYSA-N
Synonym 1,2,3-thiadiazole-4-carboxylic acid, 1,2,3 thiadiazole-4-carboxylic acid, thiadiazolecarboxylicacid, 4-carboxy-1,2,3-thiadiazole, 1,2,3-thiadiazole4-carboxylic acid, 1 pound not2 pound not3 thiadiazole-4-carboxylicacid
PubChem CID 351418
IUPAC Name thiadiazole-4-carboxylic acid
SMILES C1=C(N=NS1)C(=O)O

Specifications

Melting Point 227°C to 228°C
Quantity 1 g
Beilstein 113707
Solubility Information Slightly soluble in water.
Formula Weight 130.12
Percent Purity 97%
Chemical Name or Material 1,2,3-Thiadiazole-4-carboxylic acid

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.