missing translation for 'onlineSavingsMsg'
Learn More

1,2,3-Trichloropropane, 98+%

Product Code. 11386886
Change view
Click to view available options
Quantity:
250 g
1000 g
Unit Size:
1kg
250g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11386886 250 g 250g
11396886 1000 g 1kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11386886 Supplier Thermo Scientific Alfa Aesar Supplier No. L04312.30
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 96-18-4 | C3H5Cl3 | 147.42 g/mol

1,2,3-Trichloropropane is used to study its reaction with OH regarding it spotential atmospheric relevance.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1,2,3-Trichloropropane is used to study its reaction with OH regarding it spotential atmospheric relevance.

Solubility
Slightly soluble in water (2 g/L at 25°C).

Notes
Special handling precautions required. View MSDS prior to purchase. MSDS are available online at www.alfa.com
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 96-18-4
Molecular Formula C3H5Cl3
Molecular Weight (g/mol) 147.42
MDL Number MFCD00000946
InChI Key CFXQEHVMCRXUSD-UHFFFAOYSA-N
Synonym trichlorohydrin, allyl trichloride, propane, 1,2,3-trichloro, glycerol trichlorohydrin, glyceryl trichlorohydrin, unii-3mj7qck0z0, ccris 5874, 3mj7qck0z0, dsstox_cid_1390, dsstox_rid_76132
PubChem CID 7285
ChEBI CHEBI:34036
IUPAC Name 1,2,3-trichloropropane
SMILES ClCC(Cl)CCl

Specifications

Melting Point -14°C
Density 1.385
Boiling Point 156°C to 158°C
Flash Point 74°C (165°F)
Refractive Index 1.4822
Quantity 250 g
UN Number UN2810
Beilstein 1732068
Solubility Information Slightly soluble in water (2g/L at 25°C).
Formula Weight 147.43
Percent Purity ≥98%
Chemical Name or Material 1,2,3-Trichloropropane
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • 1,2,3-trichloropropane
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • Carcinogenicity Category 1B
  • Reproductive toxicity Category 1B
Hazard Statement
  • H302-Harmful if swallowed.
  • H312-Harmful in contact with skin.
  • H332-Harmful if inhaled.
  • H350-May cause cancer.
  • H360F-May damage fertility.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.
Supplemental information
  • MIXTURE LIST-Contain: 1,2,3-Trichloropropane

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.