missing translation for 'onlineSavingsMsg'
Learn More

1,2,4,5-Tetramethylbenzene, 97+%

Product Code. 15557029 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
100 g
500 g
Unit Size:
100g
500g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15557029 500 g 500g
15547029 5 g 5g
15502245 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15557029 Supplier Thermo Scientific Acros Supplier No. 138375000
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 95-93-2 | C10H14 | 134.22 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 95-93-2
Molecular Formula C10H14
Molecular Weight (g/mol) 134.22
InChI Key SQNZJJAZBFDUTD-UHFFFAOYSA-N
Synonym durene, durol, benzene, 1,2,4,5-tetramethyl, unii-181426cfyb, p-xylene, 2,5-dimethyl, dsstox_cid_9124, dsstox_rid_78675, dsstox_gsid_29124, duren
PubChem CID 7269
ChEBI CHEBI:38978
IUPAC Name 1,2,4,5-tetramethylbenzene
SMILES CC1=CC(=C(C=C1C)C)C

Specifications

Melting Point 78.0°C to 82.0°C
Boiling Point 196.0°C to 197.0°C
Flash Point 73°C
Infrared Spectrum Authentic
Assay Percent Range 97% min. (GC)
Linear Formula C6H2(CH3)4
Quantity 500 g
Beilstein 05,431
Merck Index 15,3515
Solubility Information Solubility in water: insoluble. Other solubilities: soluble in alcohol, ether and benzene
Formula Weight 134.22
Percent Purity 97+%
Chemical Name or Material 1,2,4,5-Tetramethylbenzene
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.