missing translation for 'onlineSavingsMsg'
Learn More

1,2,4,5-Tetramethylbenzene, 97+%

Product Code. 11351607
Change view
Click to view available options
Quantity:
100 g
500 g
Unit Size:
100g
500g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11351607 100 g 100g
11361607 500 g 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11351607 Supplier Thermo Scientific Alfa Aesar Supplier No. L04493.22
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 95-93-2 | C10H14 | 134.222 g/mol

1,2,4,5-Tetramethylbenzene is the precursor to pyromellitic dianhydride, which is used for manufacturing curing agents, adhesives, coating materials. It is used in the manufacture of some raw materials for engineering plastics (polyimides) and cross-linking agent for alkyd resins.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1,2,4,5-Tetramethylbenzene is the precursor to pyromellitic dianhydride, which is used for manufacturing curing agents, adhesives, coating materials. It is used in the manufacture of some raw materials for engineering plastics (polyimides) and cross-linking agent for alkyd resins.

Solubility
Soluble in alcohol, ether, benzene, and acetone. Insoluble in water.

Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 95-93-2
Molecular Formula C10H14
Molecular Weight (g/mol) 134.222
MDL Number MFCD00008528
InChI Key SQNZJJAZBFDUTD-UHFFFAOYSA-N
Synonym durene, durol, benzene, 1,2,4,5-tetramethyl, unii-181426cfyb, p-xylene, 2,5-dimethyl, dsstox_cid_9124, dsstox_rid_78675, dsstox_gsid_29124, duren
PubChem CID 7269
ChEBI CHEBI:38978
IUPAC Name 1,2,4,5-tetramethylbenzene
SMILES CC1=CC(=C(C=C1C)C)C

Specifications

Melting Point 77°C to 80°C
Boiling Point 191°C to 193°C
Flash Point 73°C (163°F)
Quantity 100 g
UN Number UN1325
Beilstein 1903393
Merck Index 14,3468
Solubility Information Soluble in alcohol,ether,benzene,and acetone. Insoluble in water.
Formula Weight 134.22
Percent Purity ≥97%
Chemical Name or Material 1,2,4,5-Tetramethylbenzene
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.