missing translation for 'onlineSavingsMsg'
Learn More

1,2,4-Benzenetricarboxylic Acid 1,2-Dibutyl Ester, TRC

Product Code. 30339499
Change view
Click to view available options
Quantity:
250 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30339499 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30339499 Supplier Toronto Research Chemicals Supplier No. TRCB190320250MG
 Restricted Item
Estimated Shipment: 16-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 1,2,4-Benzenetricarboxylic Acid 1,2-Dibutyl Ester
CAS 165044-69-9
Molecular Formula C17H22O6
SMILES CCCCOC(=O)c1ccc(cc1C(=O)OCCCC)C(=O)O
Molecular Weight (g/mol) 322.35
Formula Weight 322.1416 g/mol
Percent Purity >95% (HPLC)
Quantity 250 mg
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.