missing translation for 'onlineSavingsMsg'
Learn More

1,2,4-Benzenetricarboxylic acid, 98%

Product Code. p-3756869 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
50 g
250 g
Packaging:
Glass Bottle
Plastic Bottle
Unit Size:
250g
50g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10717782 5 g Glass Bottle 5g
10153581 50 g Plastic Bottle 50g
10732305 250 g Plastic Bottle 250g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10717782 Supplier Thermo Scientific Acros Supplier No. 349030050
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 528-44-9 | C9H6O6 | 210.14 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 528-44-9
Molecular Formula C9H6O6
Molecular Weight (g/mol) 210.14
MDL Number MFCD00002470
InChI Key ARCGXLSVLAOJQL-UHFFFAOYSA-N
Synonym 1,2,4-benzenetricarboxylic acid, trimellitic acid, 1,2,4-tricarboxybenzene, 4-carboxyphthalic acid, 1,3,4-benzenetricarboxylic acid, 1,4,5-benzenetricarboxylic acid, 1,2,4-benzenetricarboxylicacid, unii-7nvy29mq5f, 7nvy29mq5f, dsstox_cid_1487
PubChem CID 10708
ChEBI CHEBI:166055
IUPAC Name benzene-1,2,4-tricarboxylic acid
SMILES OC(=O)C1=CC=C(C(O)=O)C(=C1)C(O)=O

Specifications

CAS Min % 97.5
CAS Max % 100.0
Color White
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Glass Bottle
Linear Formula C6H3(COH)3
Quantity 5 g
Beilstein 09, 977
Merck Index 15, 9871
Formula Weight 210.14
Percent Purity 98%
Physical Form Crystalline Powder
Chemical Name or Material 1, 2, 4-Benzenetricarboxylic acid
Show More Show Less
compliance-icons
Product Identifier
  • 1,2,4-Benzenetricarboxylic acid
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.