missing translation for 'onlineSavingsMsg'
Learn More

1,2,4-Benzenetricarboxylic anhydride, 97%

Product Code. 11474240
Change view
Click to view available options
Quantity:
250 g
1 kg
Unit Size:
1kg
250g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11474240 1 kg 1kg
11416176 250 g 250g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11474240 Supplier Thermo Scientific Alfa Aesar Supplier No. B23003.A1
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 552-30-7 | C9H4O5 | 192.126 g/mol

1,2,4-Benzenetricarboxylic anhydride is used to produce good heat-proof, weather-proof and solvent-proof trimellitate plasticizers, to synthesize polyester resins, curing agent of epoxy resins, heat-proof insulated laminas, heat-proof varnishes, stabilizers, dyestuffs, pigments, surfactants. TMA can also be used as accelerant for vulcanization, treating agent of fiber and flocculant of film. It has wide application to high class aerooil, impregnating oil for power capacitors, adhesives for granules, instant adhesives, sizing agents, antismoke agents, dyestuffs, printing inks, medicines and farm chemicals.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1,2,4-Benzenetricarboxylic anhydride is used to produce good heat-proof, weather-proof and solvent-proof trimellitate plasticizers, to synthesize polyester resins, curing agent of epoxy resins, heat-proof insulated laminas, heat-proof varnishes, stabilizers, dyestuffs, pigments, surfactants. TMA can also be used as accelerant for vulcanization, treating agent of fiber and flocculant of film. It has wide application to high class aerooil, impregnating oil for power capacitors, adhesives for granules, instant adhesives, sizing agents, antismoke agents, dyestuffs, printing inks, medicines and farm chemicals.

Solubility
Hydrolyzes in water. Soluble in ethanol, acetone, and most other organic solvents.

Notes
Moisture sensitive. Store under inert gas. Incompatible with oxidizing agents, bases, water/ moisture.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 552-30-7
Molecular Formula C9H4O5
Molecular Weight (g/mol) 192.126
MDL Number MFCD00005925
InChI Key SRPWOOOHEPICQU-UHFFFAOYSA-N
Synonym trimellitic anhydride, 1,2,4-benzenetricarboxylic anhydride, anhydrotrimellic acid, trimellitic acid anhydride, 4-carboxyphthalic anhydride, tman, anhydrotrimellitic acid, trimellic acid anhydride, 1,3-dioxo-1,3-dihydroisobenzofuran-5-carboxylic acid, trimellitic acid 1,2-anhydride
PubChem CID 11089
ChEBI CHEBI:53050
IUPAC Name 1,3-dioxo-2-benzofuran-5-carboxylic acid
SMILES C1=CC2=C(C=C1C(=O)O)C(=O)OC2=O

Specifications

Melting Point 165°C to 169°C
Density 1.54
Boiling Point 240°C to 245°C (14 mmHg)
Flash Point 227°C (440°F)
Quantity 1 kg
Beilstein 9394
Sensitivity Moisture sensitive
Merck Index 14,9703
Solubility Information Hydrolyzes in water. Soluble in ethanol,acetone,and most other organic solvents.
Formula Weight 192.13
Percent Purity 97%
Chemical Name or Material 1,2,4-Benzenetricarboxylic anhydride
Show More Show Less
compliance-icons compliance-icons compliance-icons
Product Identifier
  • 1,2,4-Benzenetricarboxylic anhydride
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Respiratory sensitiser Category 1
  • Skin sensitiser Category 1
  • Specific target organ toxicity Category 3
Hazard Statement
  • H317-May cause an allergic skin reaction.
  • H318-Causes serious eye damage.
  • H334-May cause allergy or asthma symptoms or breathing difficulties if inhaled.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P342+P311-If experiencing respiratory symptoms: Call a POISON CENTER/doctor/ .
Supplemental information
  • MIXTURE LIST-Contain: Trimellitic anhydride

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.