missing translation for 'onlineSavingsMsg'
Learn More

1-[2-(4-chlorophenyl)-6H-1,3-thiazin-5-yl]ethan-1-one, 97%, Thermo Scientific™

Product Code. 10577893
Change view
Click to view available options
Quantity:
1 g
10 g
50 g
Unit Size:
10g
1g
50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10577893 1 g 1g
10578083 10 g 10g
10118562 50 g 50g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10577893 Supplier Thermo Scientific Maybridge Supplier No. BTB05406DA
 Restricted Item
Estimated Shipment: 16-11-2026
Add to basket
This item is not returnable. View return policy

Chemical Identifiers

CAS 219539-29-4
Molecular Formula C12H10ClNOS
Molecular Weight (g/mol) 251.73
MDL Number MFCD00276113
InChI Key NEKOTKLWPGNIOI-UHFFFAOYSA-N
Synonym 1-2-4-chlorophenyl-6h-1,3-thiazin-5-yl ethan-1-one, 1-2-4-chlorophenyl-6h-1,3-thiazin-5-yl ethanone, maybridge1_001465, ethanone,1-2-4-chlorophenyl-6h-1,3-thiazin-5-yl
PubChem CID 2799112
IUPAC Name 1-[2-(4-chlorophenyl)-6H-1,3-thiazin-5-yl]ethanone
SMILES CC(=O)C1=CN=C(SC1)C1=CC=C(Cl)C=C1

Specifications

CAS Min % 96.5
CAS Max % 100.0
Quantity 1 g
Packaging Amber glass bottle
Assay Percent Range 97%
Formula Weight 251.74
Percent Purity 97%
Chemical Name or Material 1-[2-(4-chlorophenyl)-6H-1, 3-thiazin-5-yl]ethan-1-one
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.