missing translation for 'onlineSavingsMsg'
Learn More

1-(2,4-Dichlorophenyl)-1-cyclopropyl cyanide, 98%, Thermo Scientific™

Product Code. 10486950 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
25 g
Unit Size:
25g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
10486950 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10486950 Supplier Thermo Scientific Acros Supplier No. 134970250

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Specifications

CAS 71463-55-3
Melting Point 81°C to 85°C
Assay Percent Range 97.5% min. (GC)
Packaging Glass Bottle
Molecular Formula C10H7Cl2N
MDL Number MFCD00044818
Quantity 25 g
Synonym 1-2,4-dichlorophenyl cyclopropanecarbonitrile, 1-2,4-dichlorophenyl cyclopropane-1-carbonitrile, 1-2,4-dichlorophenyl-1-cyclopropyl cyanide, 1-2,4-dichlorophenyl-1-cyanocyclopropane, 1-2,4-dichloro-phenyl-cyclopropanecarbonitrile, cyclopropanecarbonitrile, 1-2,4-dichlorophenyl
InChI Key VDKKBWVLLSVWKI-UHFFFAOYSA-N
SMILES C1CC1(C#N)C2=C(C=C(C=C2)Cl)Cl
IUPAC Name 1-(2,4-dichlorophenyl)cyclopropane-1-carbonitrile
Molecular Weight (g/mol) 212.08
PubChem CID 1268258
Formula Weight 212.08
Percent Purity 98%
Chemical Name or Material 1-(2, 4-Dichlorophenyl)-1-cyclopropyl cyanide, 98%
Show More Show Less
compliance-icons
Product Identifier
  • 1-(2,4-Dichlorophenyl)-1-cyclopropyl cyanide
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 3
Hazard Statement
  • H311-Toxic in contact with skin.
  • H331-Toxic if inhaled.
  • H301-Toxic if swallowed.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.