missing translation for 'onlineSavingsMsg'
Learn More

1,2,4-Triazol-4-amine, TRC

Product Code. 30451540
Change view
Click to view available options
Quantity:
1 g
10 g
Unit Size:
1g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30451540 1 g 1g
30454986 10 g -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30451540 Supplier Toronto Research Chemicals Supplier No. TRCT7674201G
 Restricted Item
Estimated Shipment: 21-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 584-13-4
Molecular Formula C2 H4 N4
Molecular Weight (g/mol) 84.08
Synonym 4-Amino-1,2,4-triazole, 4H-1,2,4-Triazole, 4-amino- (8CI), 1,2,4-Triazol-4-amine, 1,2,4-Triazol-4-ylamine, 1,2,4-Triazole-4-amine, 1-Amino-1,3,4-triazole, 1-Amino-1H-1,3,4-triazole, 4-Amino-1,2,4(4H)-triazole, 4-Amino-4H-1,2,4-triazole, A 1137, NSC 3263, NSC 7242, 4H-1,2,4-Triazol-4-amine, ATC
IUPAC Name 1,2,4-triazol-4-amine
SMILES Nn1cnnc1

Specifications

Quantity 1 g
InChI Formula InChI=1S/C2H4N4/c3-6-1-4-5-2-6/h1-2H,3H2
Formula Weight 84.0436
Percent Purity >95%
Chemical Name or Material 4-Amino-1,2,4-triazole
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.