missing translation for 'onlineSavingsMsg'
Learn More

1,2,4-Triazole Sodium Salt, TRC

Product Code. 30454660
Change view
Click to view available options
Quantity:
25 g
Unit Size:
25g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30454660 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30454660 Supplier Toronto Research Chemicals Supplier No. TRCT76744025G
 Restricted Item
Estimated Shipment: 01-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 1,2,4-Triazole Sodium Salt
CAS 41253-21-8
Molecular Formula C2H2N3Na
Quantity 25 g
Synonym 1H-1,2,4-Triazole Sodium Salt, Sodium 1,2,4-triazole-4-ide, Sodium 1H-1,2,4-Triazolate, Sodium 1H-1,2,4-Triazolide, Triazole Sodium Salt
InChI Formula InChI=1S/C2H2N3.Na/c1-3-2-5-4-1;/h1-2H;/q-1;+1
SMILES [Na+].c1nc[n-]n1
IUPAC Name sodium;2,4-diaza-1-azanidacyclopenta-2,4-diene
Molecular Weight (g/mol) 91.05
Formula Weight 91.0146
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.