missing translation for 'onlineSavingsMsg'
Learn More

1,2,4-Trichlorobenzene, Packaged under Argon, HPLC Grade, J.T. Baker™

Product Code. 15568614 Shop All J.T.Baker Products
Change view
Click to view available options
Quantity:
3.8 L
Packaging:
Narrow Mouth PVC Coated Amber Glass Bottle
Unit Size:
3.8 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
15568614 3.8 L Narrow Mouth PVC Coated Amber Glass Bottle 3.8 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Regulatory: This product contains a substance listed in Annex XVII to REACH Regulation (EC) No 1907/2006, all customers must complete a declaration before delivery.
Product Code. 15568614 Supplier Avantor J.T.Baker Supplier No. 944405
  • ONLINE EXCLUSIVE PRICE
    550.61 €
    479.99 € / 3.8 L
    Save 70.62 €
    12% Off
 Restricted Item
Estimated Shipment: 26-05-2027
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy
Regulatory: This product contains a substance listed in Annex XVII to REACH Regulation (EC) No 1907/2006, all customers must complete a declaration before delivery.

High purity solvents for use in analytical and preparative separations

    Suitable for use in gel permeation chromatography

Specifications

CAS 120-82-1
Quantity 3.8 L
Packaging Narrow Mouth PVC Coated Amber Glass Bottle
Specific Gravity 1L = 1.45kg
Molecular Formula C6H3Cl3
Synonym benzene, 1,2,4-trichloro, unsym-trichlorobenzene, hostetex l-pec, trojchlorobenzen, 1,2,4-trichlorbenzol, 1,2,4-trichlorobenzol, 1,3,4-trichlorobenzene, trichlorobenzene a, 1,2,5-trichlorobenzene, as-trichlorobenzene
InChI Key PBKONEOXTCPAFI-UHFFFAOYSA-N
SMILES C1=CC(=C(C=C1Cl)Cl)Cl
IUPAC Name 1,2,4-trichlorobenzene
Molecular Weight (g/mol) 181.44
PubChem CID 13
ChEBI CHEBI:28222
Formula Weight 181.45
Grade HPLC
Chemical Name or Material 1,2,4-Trichlorobenzene, Packaged under Argon
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.