missing translation for 'onlineSavingsMsg'
Learn More

1-(2,5-Dimethylphenyl)ethanol, TRC

Product Code. 30380093
Change view
Click to view available options
Quantity:
100 mg
500 mg
1 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30380093 500 mg
30373649 100 mg
30375567 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30380093 Supplier Toronto Research Chemicals Supplier No. TRCD460198500MG
 Restricted Item
Estimated Shipment: 14-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 32917-52-5
Molecular Formula C10 H14 O
Molecular Weight (g/mol) 150.22
Synonym α,2,5-Trimethylbenzenemethanol, 1-(2,5-Dimethylphenyl)ethan-1-ol, 2,5-Dimethylphenyl-methyl-carbinol, α,2,5-Trimethylbenzyl Alcohol
IUPAC Name 1-(2,5-dimethylphenyl)ethanol
SMILES CC(O)c1cc(C)ccc1C

Specifications

Quantity 500 mg
InChI Formula InChI=1S/C10H14O/c1-7-4-5-8(2)10(6-7)9(3)11/h4-6,9,11H,1-3H3
Formula Weight 150.1045
Chemical Name or Material 1-(2,5-Dimethylphenyl)ethanol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.