missing translation for 'onlineSavingsMsg'
Learn More

1-(2,5-Dimethylphenyl)piperazine, 96%, Thermo Scientific™

Product Code. 11347585
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11347585 25 g 25g
11337585 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11347585 Supplier Thermo Scientific Alfa Aesar Supplier No. L05744.14
Request Bulk or Custom Format

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1013-25-8
Molecular Formula C12H19N2
Molecular Weight (g/mol) 191.30
MDL Number MFCD00038378
InChI Key YRIFWVMRUFKWLM-UHFFFAOYSA-O
Synonym 1-2,5-dimethylphenyl piperazine, 1-2,5-xylyl piperazine, piperazine, 1-2,5-dimethylphenyl, 2,5-dimethylphenyl piperazine, acmc-1bpa9, 2,5-dimethylphenylpiperazine, n-2,5-dimethylphenyl piperazine, 1-2,5-dimethylphenyl-piperazine, 4-2,5-dimethyl-phenyl piperazine, 1-2,5-dimethyl-phenyl-piperazine
PubChem CID 70542
IUPAC Name 1-(2,5-dimethylphenyl)piperazine
SMILES CC1=CC=C(C)C(=C1)N1CC[NH2+]CC1

Specifications

Melting Point 42°C to 47°C
Quantity 25 g
UN Number UN3259
Beilstein 611229
Boiling Point 154°C (10 mmHg)
Formula Weight 190.28
Percent Purity 96%
Chemical Name or Material 1-(2,5-Dimethylphenyl)piperazine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.