missing translation for 'onlineSavingsMsg'
Learn More

1,2,6-Hexanetriol, 97+%, extra pure

Product Code. 10447150 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
100 g
500 g
Packaging:
Glass bottle
Unit Size:
100g
500g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10447150 500 g Glass bottle 500g
10265140 100 g Glass bottle 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10447150 Supplier Thermo Scientific Acros Supplier No. 139835000
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 106-69-4 | C6H14O3 | 134.18 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 106-69-4
Molecular Formula C6H14O3
Molecular Weight (g/mol) 134.18
MDL Number MFCD00002976
InChI Key ZWVMLYRJXORSEP-UHFFFAOYNA-N
Synonym 1,2,6-hexanetriol, 1,2,6-trihydroxyhexane, hexanetriol-1,2,6, 1,6-hexanetriol, hexanetriol-1,6, hexane-1,6-triol, 1,6-trihydroxyhexane, acmc-1bsub, dsstox_cid_21224
PubChem CID 7823
IUPAC Name hexane-1,2,6-triol
SMILES OCCCCC(O)CO

Specifications

CAS Min % 97.0
CAS Max % 100.0
Melting Point 25.0°C to 32.0°C
Color Yellow
Density 1.1090g/mL
pH 4.0 to 7.0 (30% soln. at 20°C)
Boiling Point 178.0°C (6.7 mmHg)
Flash Point 198°C
Infrared Spectrum Authentic
Assay Percent Range 97% min. (GC)
Packaging Glass bottle
Linear Formula HO(CH2)4CH(OH)CH2OH
Refractive Index 1.4750 to 1.4770
Quantity 500 g
Beilstein 01,IV,2784
Specific Gravity 1.109
Solubility Information Solubility in water: miscible with water. Other solubilities: miscible with acetone and alcohol, immiscible with benzene, toluene, heptane and, diethyl ether, practically insoluble in ether, low solubility in non-polar solvents
Formula Weight 134.18
Percent Purity 97+%
Grade Extra Pure
Physical Form Viscous Liquid
Chemical Name or Material 1, 2, 6-Hexanetriol, 97%
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.