missing translation for 'onlineSavingsMsg'
Learn More

1-(2-Aminoethyl)piperazine, TRC

Product Code. 30134304
Change view
Click to view available options
Quantity:
10 g
100 g
Unit Size:
100g
10g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30134304 100 g 100g
30006365 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30134304 Supplier Toronto Research Chemicals Supplier No. TRCA609255100G
 Restricted Item
Estimated Shipment: 25-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 140-31-8
Molecular Formula C6 H15 N3
Molecular Weight (g/mol) 129.2034
Synonym 1-Piperazineethanamine, Piperazine, 1-(2-aminoethyl)- (6CI,7CI,8CI), 1-(2-Aminoethyl)piperazine, 1-(β-Aminoethyl)piperazine, 1-Piperazineethylamine, 2-(1-Piperazinyl)ethanamine, 2-(1-Piperazinyl)ethylamine, 2-(Piperazin-1-yl)ethaneamine, 2-(Piperazinyl-1-yl)ethanamine, 2-Piperazinylethylamine, 4-(2-Aminoethyl)piperazine, AEP, Ancamine AEP, Epicure 3200, Epikure 3202, N-(β-Aminoethyl)piperazine, N-AEP, NSC 38968
IUPAC Name 2-piperazin-1-ylethanamine
SMILES NCCN1CCNCC1

Specifications

Quantity 100 g
InChI Formula InChI=1S/C6H15N3/c7-1-4-9-5-2-8-3-6-9/h8H,1-7H2
Formula Weight 129.1266
Chemical Name or Material N-(2-Aminoethyl)piperazine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.