missing translation for 'onlineSavingsMsg'
Learn More

1-(2-Aminoethyl)piperidine, 98%

Product Code. 11448297
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11448297 5 g 5g
11458297 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11448297 Supplier Thermo Scientific Alfa Aesar Supplier No. A18373.06
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 27578-60-5 | C7H16N2 | 128.219 g/mol

1-(2-Aminoethyl)piperidine is used in modification of vinylbenzyl chloride/divinylbenzene gel copolymer bead and in the synthesis of linkage isomers trans-bis[1-(2-aminoethyl)piperidine]dinitronickel and trans-bis[1-(2-aminoethyl)-piperidine]dinitritonickel. It is also used as a reactant for synthesis of: analogs of anticancer agents, inhibitor of botulinum neurotoxin serotype A light chain, P. falciparum malaria, and Ebola filovirus, cannabinoid CB1 receptor antagonists, small molecules that restore E-cadherin expression and reduce invasion in colorectal carcinoma cells, potent and selective 5-HT6 antagonists and N-mustards as anticancer agents. 1-(2-Aminoethyl)piperidine on [1+1] condensation reaction with 3-methoxy salicylaldehyde yields tridentate Schiff base ligand.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1-(2-Aminoethyl)piperidine is used in modification of vinylbenzyl chloride/divinylbenzene gel copolymer bead and in the synthesis of linkage isomers trans-bis[1-(2-aminoethyl)piperidine]dinitronickel and trans-bis[1-(2-aminoethyl)-piperidine]dinitritonickel. It is also used as a reactant for synthesis of: analogs of anticancer agents, inhibitor of botulinum neurotoxin serotype A light chain, P. falciparum malaria, and Ebola filovirus, cannabinoid CB1 receptor antagonists, small molecules that restore E-cadherin expression and reduce invasion in colorectal carcinoma cells, potent and selective 5-HT6 antagonists and N-mustards as anticancer agents. 1-(2-Aminoethyl)piperidine on [1+1] condensation reaction with 3-methoxy salicylaldehyde yields tridentate Schiff base ligand.

Solubility
Partly miscible in water.

Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents, acid chlorides and acid anhydrides. It is sensitive to air.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 27578-60-5
Molecular Formula C7H16N2
Molecular Weight (g/mol) 128.219
MDL Number MFCD00006516
InChI Key CJNRGSHEMCMUOE-UHFFFAOYSA-N
Synonym n-2-aminoethyl piperidine, 1-2-aminoethyl piperidine, 2-piperidin-1-yl ethanamine, 2-piperidin-1-yl ethan-1-amine, 1-piperidineethanamine, 2-piperidinoethylamine, 2-1-piperidinyl ethanamine, 2-1-piperidinyl ethylamine, n-aminoethylpiperidine, 2-piperidino-1-ethanamine
PubChem CID 33944
IUPAC Name 2-piperidin-1-ylethanamine
SMILES C1CCN(CC1)CCN

Specifications

Density 0.903
Boiling Point 185°C to 186°C
Flash Point 57°C (134°F)
Refractive Index 1.474
Quantity 5 g
UN Number UN2733
Beilstein 103469
Sensitivity Air Sensitive
Solubility Information Partly miscible in water.
Formula Weight 128.22
Percent Purity 98%
Chemical Name or Material 1-(2-Aminoethyl)piperidine
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.