missing translation for 'onlineSavingsMsg'
Learn More

1,2-Bis(2,4,6-tribromophenoxy)ethane-13C12, TRC

Product Code. 30340406
Change view
Click to view available options
Quantity:
5 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30340406 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30340406 Supplier Toronto Research Chemicals Supplier No. TRCB5854285MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 1,2-Bis(2,4,6-tribromophenoxy)ethane-13C12
Molecular Formula 13C12 C2 H4 Br6 O2
Quantity 5 mg
Synonym 1,1'-[1,2-Ethanediylbis(oxy)]bis[2,4,6-tribromobenzene]-13C12
InChI Formula InChI=1S/C14H4Br6O2/c15-7-3-9(17)13(10(18)4-7)21-1-2-22-14-11(19)5-8(16)6-12(14)20/h1-2H2/i3+1,4+1,5+1,6+1,7+1,8+1,9+1,10+1,11+1,12+1,13+1,14+1
SMILES Br[13c]1[13c][13c](Br)[13c](OCCO[13c]2[13c](Br)[13c][13c](Br)[13c][13c]2Br)[13c](Br)[13c]1
IUPAC Name 1,3,5-tribromo-2-[2-(2,4,6-tribromo(1,2,3,4,5,6-13C6)cyclohexa-1,3,5-trien-1-yl)oxyethoxy](1,2,3,4,5,6-13C6)cyclohexa-1,3,5-triene
Molecular Weight (g/mol) 695.516
Formula Weight 689.571 g/mol
Percent Purity >95% (HPLC)
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.