missing translation for 'onlineSavingsMsg'
Learn More

1,2-Bis[2-(4-Dibenzo[b,f][1,4]thiazepin-11-yl-1-piperazinyl)ethoxy]ethane, TRC

Product Code. 30136928
Change view
Click to view available options
Quantity:
10 mg
100 mg
Unit Size:
100mg
10mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30136928 100 mg 100mg
30008490 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30136928 Supplier Toronto Research Chemicals Supplier No. TRCB419120100MG
 Restricted Item
Estimated Shipment: 19-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1371638-05-9
Molecular Formula C40H44N6O2S2
Molecular Weight (g/mol) 704.95
Synonym Quetiapine EP Impurity E;
IUPAC Name 6-[4-[2-[2-[2-(4-benzo[b][1,4]benzothiazepin-6-ylpiperazin-1-yl)ethoxy]ethoxy]ethyl]piperazin-1-yl]benzo[b][1,4]benzothiazepine
SMILES C(COCCN1CCN(CC1)C2=Nc3ccccc3Sc4ccccc24)OCCN5CCN(CC5)C6=Nc7ccccc7Sc8ccccc68

Specifications

Quantity 100 mg
InChI Formula InChI=1S/C40H44N6O2S2/c1-5-13-35-31(9-1)39(41-33-11-3-7-15-37(33)49-35)45-21-17-43(18-22-45)25-27-47-29-30-48-28-26-44-19-23-46(24-20-44)40-32-10-2-6-14-36(32)50-38-16-8-4-12-34(38)42-40/h1-16H,17-30H2
Formula Weight 704.2967
Chemical Name or Material 1,2-Bis[2-(4-Dibenzo[b,f][1,4]thiazepin-11-yl-1-piperazinyl)ethoxy]ethane
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.