missing translation for 'onlineSavingsMsg'
Learn More

1,2-Bis(2-aminophenoxy)-ethane-N,N,N'N'-tetraacetic acid, 97%

Product Code. 10433025 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
100 mg
1 g
5 g
Packaging:
Glass Bottle
Unit Size:
100mg
1g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10433025 5 g Glass Bottle 5g
10360984 100 mg Glass Bottle 100mg
10408280 1 g Glass Bottle 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10433025 Supplier Thermo Scientific Acros Supplier No. 123310050
 Restricted Item
Estimated Shipment: 30-09-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 85233-19-8 | C22H24N2O10 | 476.44 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chelation/Complexation reagent
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 85233-19-8
Molecular Formula C22H24N2O10
Molecular Weight (g/mol) 476.44
MDL Number MFCD00036255
InChI Key FTEDXVNDVHYDQW-UHFFFAOYSA-N
Synonym bapta, bapeta, 1,2-bis 2-aminophenoxy ethane-n,n,n',n'-tetraacetic acid, ca ii-bapta, 1,2-bis-2-aminophenoxy ethane n,n,n',n'-tetraacetic acid, unii-k22ddw77c0, 1,2-bis o-aminophenoxy ethane-n,n,n',n'-tetraacetic acid, bapta tetrasodium salt hydrate, glycine, n,n'-1,2-ethanediylbis oxy-2,1-phenylene bis n-carboxymethyl, 2,2',2,2'-ethane-1,2-diylbis oxy-2,1-phenylenenitrilo tetraacetic acid
PubChem CID 104751
ChEBI CHEBI:60888
IUPAC Name 2-{[2-(2-{2-[bis(carboxymethyl)amino]phenoxy}ethoxy)phenyl](carboxymethyl)amino}acetic acid
SMILES OC(=O)CN(CC(O)=O)C1=CC=CC=C1OCCOC1=CC=CC=C1N(CC(O)=O)CC(O)=O

Specifications

CAS Min % 96.0
CAS Max % 100.0
Melting Point 177.0°C to 179.0°C
Color White
Infrared Spectrum Authentic
Assay Percent Range 97%
Packaging Glass Bottle
Quantity 5 g
Merck Index 15, 952
Solubility Information Solubility in water: soluble.
Formula Weight 476.44
Percent Purity 97%
Physical Form Powder
Chemical Name or Material 1,2-Bis(2-aminophenoxy)-ethane-N,N,N'N'-tetraacetic acid
Show More Show Less
compliance-icons
Product Identifier
  • 1,2-Bis(2-aminophenoxy)-ethane-N,N,N'N'-tetraacetic acid
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.