missing translation for 'onlineSavingsMsg'
Learn More

1,2-Bis(2-chloroethoxy)ethane, 97%, ACROS Organics™

Product Code. 10479052 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 mL
Unit Size:
5mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10479052 5 mL 5mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10479052 Supplier Thermo Scientific Acros Supplier No. 349620050
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Chemical Identifiers

CAS 112-26-5
Molecular Formula C6H12Cl2O2
Molecular Weight (g/mol) 187.06
MDL Number MFCD00000976
InChI Key AGYUOJIYYGGHKV-UHFFFAOYSA-N
Synonym 1,2-bis 2-chloroethoxy ethane, triglycol dichloride, triethylene glycol dichloride, ethane, 1,2-bis 2-chloroethoxy, 1,8-dichloro-3,6-dioxaoctane, 1,2-bis chloroethoxy ethane, bis 2-chloroethoxy ethane, di 2-chloroethyl cellosolve, 2-2-chloroethoxy ethyl 2'-chloroethyl ether, unii-28jq82m13j
PubChem CID 8171
IUPAC Name 1-chloro-2-[2-(2-chloroethoxy)ethoxy]ethane
SMILES ClCCOCCOCCCl

Specifications

CAS Min % 96.0
CAS Max % 100.0
Color Colorless to Yellow
Infrared Spectrum Authentic
Assay Percent Range 97%
Packaging Glass bottle
Linear Formula (-CH2OCH2CH2Cl)2
Refractive Index 1.4600 to 1.4620 (20°C, 589nm)
Quantity 5 mL
Formula Weight 187.07
Percent Purity ≥96.0%
Physical Form Liquid
Chemical Name or Material 1, 2-Bis(2-chloroethoxy)ethane, 97%
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.