missing translation for 'onlineSavingsMsg'
Learn More

(-)-1,2-Bis[(2R,5R)-2,5-diethyl-1-phospholanyl]benzene, 97+%

Product Code. 15495776
Change view
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15495776 250 mg 250mg
15485776 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15495776 Supplier Thermo Scientific Alfa Aesar Supplier No. H60470.MD
 Restricted Item
Request Bulk or Custom Format

This item requires a price update prior to being ordered.

Request a Price for this item here.

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 136705-64-1 | C22H36P2 | 362.478 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Kemiske identifikatorer

CAS 136705-64-1
Molecular Formula C22H36P2
Molecular Weight (g/mol) 362.478
MDL Number MFCD00142335
InChI Key GVVCHDNSTMEUCS-UAFMIMERSA-N
Synonym ethyl-duphos, r,r, r,r-ethyl-duphos, unii-7n2eb15paq, --1,2-bis 2r,5r-2,5-diethylphospholano benzene, r,r-et-duphos, 7n2eb15paq, --duphos, r,r-et-duphos mi, 2r,5r-1-2-2r,5r-2,5-diethylphospholan-1-yl phenyl-2,5-diethylphospholane
PubChem CID 2734549
IUPAC Name (2R,5R)-1-[2-[(2R,5R)-2,5-diethylphospholan-1-yl]phenyl]-2,5-diethylphospholane
SMILES CCC1CCC(P1C2=CC=CC=C2P3C(CCC3CC)CC)CC

Tekniske data

Density 1.01
Flash Point >110°C (230°F)
Refractive Index 1.6
Quantity 250 mg
Beilstein 4814174
Sensitivity Air Sensitive
Optical Rotation −265° (c=1 in Hexane)
Formula Weight 362.47
Percent Purity ≥97%
Chemical Name or Material (-)-1,2-Bis[(2R,5R)-2,5-diethyl-1-phospholanyl]benzene

RUO – Research Use Only

Produkttitel
Vælg et problem

Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.