missing translation for 'onlineSavingsMsg'
Learn More

(+)-1,2-Bis[(2R,5R)-2,5-diethyl-1-phospholanyl]ethane, 97+%

Product Code. 15465756
Change view
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15465756 250 mg 250mg
15455756 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15465756 Supplier Thermo Scientific Alfa Aesar Supplier No. H60447.MD
Request Bulk or Custom Format

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 136705-62-9 | C18H36P2 | 314.434 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 136705-62-9
Molecular Formula C18H36P2
Molecular Weight (g/mol) 314.434
MDL Number MFCD01073768
InChI Key QOLRLVPABLMMKI-BRSBDYLESA-N
Synonym et-bpe, r,r, r,r-et-bpe, +-1,2-bis 2r,5r-2,5-diethylphospholano ethane, r,r-et-bpe mi, bpe ligands r,r-et-bpe mi, phospholane, 1,1'-1,2-ethanediyl bis 2,5-diethyl-, 2r,2'r,5r,5'r, 1,2-bis 2r,5r-2,5-diethylphospholan-1-yl ethane, 1,1'-ethylenebis 2alpha,5beta-diethyl-1-phosphacyclopentane
PubChem CID 11433485
IUPAC Name (2R,5R)-1-[2-[(2R,5R)-2,5-diethylphospholan-1-yl]ethyl]-2,5-diethylphospholane
SMILES CCC1CCC(P1CCP2C(CCC2CC)CC)CC

Specifications

Density 0.939
Boiling Point 104°C to 106°C (0.05 mmHg)
Flash Point 110°C (230°F)
Refractive Index 1.525
Quantity 250 mg
UN Number UN2845
Sensitivity Air Sensitive
Optical Rotation +320° (c=1 in Hexane)
Formula Weight 314.43
Percent Purity ≥97%
Chemical Name or Material (+)-1,2-Bis[(2R,5R)-2,5-diethyl-1-phospholanyl]ethane
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.