missing translation for 'onlineSavingsMsg'
Learn More

1,2-Bis((2R,5R)-2,5-diethylphospholano)benzene(cyclooctadiene)rhodium(I) tetrafluoroborate, 97%, Thermo Scientific Chemicals

Product Code. 10535490 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
100 mg
250 mg
Unit Size:
100mg
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
10535490 250 mg 250mg
10812261 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10535490 Supplier Thermo Scientific Acros Supplier No. 435182500
 Restricted Item
Estimated Shipment: 11-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 228121-39-9 | Molecular Formula: C30H48BF4P2Rh | Molecular Weight: 660.37 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 228121-39-9
Molecular Formula C30H48BF4P2Rh
Molecular Weight (g/mol) 660.37
MDL Number MFCD01862464
InChI Key DSYBEQKPSKLNMC-RTXTXWNLNA-N
PubChem CID 11549211
IUPAC Name cyclooctane;(2R,5R)-1-[2-[(2R,5R)-2,5-diethylphospholan-1-yl]phenyl]-2,5-diethylphospholane;rhodium;tetrafluoroborate
SMILES [Rh+].F[B-](F)(F)F.C1C\C=C/CC\C=C/1.CC[C@@H]1CC[C@@H](CC)P1C1=C(C=CC=C1)P1[C@H](CC)CC[C@H]1CC

Specifications

CAS Min % 96.0
CAS Max % 100.0
Color Brown to Orange
Elemental Analysis 52.4 to 56.8% C
Infrared Spectrum Authentic
Assay Percent Range 97%
Quantity 250 mg
Formula Weight 660.37
Percent Purity 97%
Physical Form Powder
Chemical Name or Material 1,2-Bis((2R,5R)-2,5-Diethylphospholano)benzene(cyclooctadiene)rhodium(I) tetrafluoroborate
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.