missing translation for 'onlineSavingsMsg'
Learn More

1,2-Bis(4-(benzo[d]oxazol-2-yl)phenyl)ethene, TRC

Product Code. 30348397
Change view
Click to view available options
Quantity:
500 mg
Unit Size:
500mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30348397 500 mg 500mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30348397 Supplier Toronto Research Chemicals Supplier No. TRCB410525500MG
 Restricted Item
Estimated Shipment: 09-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 4,4'-Bis(2-benzoxazolyl)stilbene
CAS 1533-45-5
Molecular Formula C28 H18 N2 O2
Quantity 500 mg
Synonym Benzoxazole, 2,2'-(vinylenedi-p-phenylene)bis- (7CI,8CI), 2,2'-(1,2-Ethenediyldi-4,1-phenylene)bis[benzoxazole], 2,2'-(Vinylenedi-p-phenylene)bisbenzoxazole, 4,4'-Bis(2-benzoxazolyl)stilbene, 4,4'-Di(benzoxazol-2-yl)stilbene, Benetex OB 1, C.I. 40674, C.I. Fluorescent Brightener 393, Eastobrite OB 1, Eastobrite OB 3, Fluorescent Brightener 393, OB 1, Optical Brightener 1, TM, TM (UV stabilizer), Uvitex OB 1, Uvitex OB-ONE, p,p'-Bis(2-benzoxazolyl)stilbene
InChI Formula InChI=1S/C28H18N2O2/c1-3-7-25-23(5-1)29-27(31-25)21-15-11-19(12-16-21)9-10-20-13-17-22(18-14-20)28-30-24-6-2-4-8-26(24)32-28/h1-18H
SMILES o1c(nc2ccccc12)c3ccc(C=Cc4ccc(cc4)c5oc6ccccc6n5)cc3
IUPAC Name 2-[4-[2-[4-(1,3-benzoxazol-2-yl)phenyl]ethenyl]phenyl]-1,3-benzoxazole
Molecular Weight (g/mol) 414.45
Formula Weight 414.1368
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.