missing translation for 'onlineSavingsMsg'
Learn More

1,2-Bis(dicyclohexylphosphino)ethane, 98%

Product Code. 11313678
Change view
Click to view available options
Quantity:
2 g
Unit Size:
2g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11313678 2 g 2g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11313678 Supplier Thermo Scientific Alfa Aesar Supplier No. 039269.04
 Restricted Item
Request Bulk or Custom Format

This item requires a price update prior to being ordered.

Request a Price for this item here.

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 23743-26-2 | C26H48P2 | 422.618 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

  • Packaged under argon
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 23743-26-2
Molecular Formula C26H48P2
Molecular Weight (g/mol) 422.618
MDL Number MFCD00015521
InChI Key BOUYBUIVMHNXQB-UHFFFAOYSA-N
Synonym 1,2-bis dicyclohexylphosphino ethane, ethylenebis dicyclohexylphosphine, phosphine, 1,2-ethanediylbis dicyclohexyl, 1,2-ethanediylbis dicyclohexyl phosphine, dicyclohexyl 2-dicyclohexylphosphanylethyl phosphane, bis 1,2-dicyclohexylphosphino ethane, pubchem6558, acmc-1capw, ethane-1,2-diylbis dicyclohexylphosphane, phosphine, 1,2-ethanediylbis*dicyclohexyl
PubChem CID 534202
IUPAC Name dicyclohexyl(2-dicyclohexylphosphanylethyl)phosphane
SMILES C1CCC(CC1)P(CCP(C2CCCCC2)C3CCCCC3)C4CCCCC4

Specifications

Quantity 2 g
UN Number UN1325
Sensitivity Air Sensitive
Formula Weight 422.61
Percent Purity 98%
Physical Form Crystalline
Chemical Name or Material 1,2-Bis(dicyclohexylphosphino)ethane

RUO – Research Use Only

Name des Produkts
Wählen Sie ein Thema

Indem Sie auf Absenden klicken, erklären Sie sich damit einverstanden, dass Fisher Scientific sich mit Ihnen in Verbindung setzen kann, um Ihr Feedback in diesem Formular zu bearbeiten. Wir werden Ihre Informationen nicht für andere Zwecke weitergeben. Alle bereitgestellten Kontaktinformationen werden in Übereinstimmung mit unserer Datenschutzrichtlinie aufbewahrt. Datenschutzrichtlinie.