missing translation for 'onlineSavingsMsg'
Learn More

1,2-Bis(dimethylsilyl)benzene, 98%

Product Code. 11340378
Change view
Click to view available options
Quantity:
0.5 g
2 g
10 g
Unit Size:
0.5g
10g
2g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11340378 2 g 2g
11330378 0.5 g 0.5g
11320378 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11340378 Supplier Thermo Scientific Alfa Aesar Supplier No. 043755.04
 Restricted Item
Estimated Shipment: 12-10-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 17985-72-7 | C10H16Si2 | 192.408 g/mol

1,2-Bis(dimethylsilyl)benzene is a protecting group-reagent for amines and amino acids forming benzostabase derivatives. It is an important raw material and in organic synthesis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 17985-72-7
Molecular Formula C10H16Si2
Molecular Weight (g/mol) 192.408
MDL Number MFCD00142462
InChI Key MUUXBTFQEXVEEI-UHFFFAOYSA-N
Synonym 1,2-bis dimethylsilyl benzene, 1,2-phenylenebis dimethylsilane, 2-dimethyl, acmc-209efy, bis dimethylsilyl benzene, amtsi084, o-phenylenebis dimethylsilane, 2-dimethylsilylphenyl-dimethylsilane, 5,6-bis dimethylsilanediyl-1,3-cyclohexadiene
PubChem CID 6364986
IUPAC Name [2-(dimethyl-$l^{3}-silanyl)phenyl]-dimethylsilicon
SMILES C[Si](C)C1=CC=CC=C1[Si](C)C

Specifications

Density 0.898 g/mL
Boiling Point 101°C (13 mmHg)
Flash Point 70°C (158°F)
Refractive Index 1.514
Quantity 2 g
Solubility Information Insoluble in water.
Formula Weight 194.43
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material 1,2-Bis (dimethylsilyl)benzene

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.