missing translation for 'onlineSavingsMsg'
Learn More

1,2-Bis(diphenylphosphino)ethane, 98+%

Product Code. 15119193 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
10 g
50 g
250 g
Packaging:
Glass bottle
Unit Size:
10g
1g
250g
50g
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
15119193 250 g Glass bottle 250g
10085373 1 g Glass bottle 1g
10736351 10 g Glass bottle 10g
10376450 50 g Glass bottle 50g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 15119193 Supplier Thermo Scientific Acros Supplier No. 147912500
 Restricted Item
Estimated Shipment: 07-09-2026
Add to basket
This item is not returnable. View return policy

CAS: 1663-45-2 | C26H24P2 | 398.43 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1663-45-2
Molecular Formula C26H24P2
Molecular Weight (g/mol) 398.43
MDL Number MFCD00003047
InChI Key QFMZQPDHXULLKC-UHFFFAOYSA-N
Synonym 1,2-bis diphenylphosphino ethane, diphos, ethylenebis diphenylphosphine, bis diphenylphosphine ethane, bis 1,2-diphenylphosphino ethane, phosphine, 1,2-ethanediylbis diphenyl, phosphine, ethylenebis diphenyl, 1,2-bis diphenylphosphine ethane, 1,2-bis diphenylphosphino-ethane, unii-kl33qe52i4
PubChem CID 74267
ChEBI CHEBI:30669
IUPAC Name 2-diphenylphosphanylethyl(diphenyl)phosphane
SMILES C(CP(C1=CC=CC=C1)C1=CC=CC=C1)P(C1=CC=CC=C1)C1=CC=CC=C1

Specifications

Melting Point 138.0°C to 142.0°C
Color White
Infrared Spectrum Authentic
Assay Percent Range 98% min. (HPLC)
Packaging Glass bottle
Linear Formula (C6H5)2PCH2CH2P(C6H5)2
Quantity 250 g
Merck Index 15,1254
Formula Weight 398.41
Percent Purity 98+%
Physical Form Crystalline Powder or Crystals
Chemical Name or Material 1, 2-Bis(diphenylphosphino)ethane, 98+%
Show More Show Less
compliance-icons
Product Identifier
  • 1,2-Bis(diphenylphosphino)ethane
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.