missing translation for 'onlineSavingsMsg'
Learn More

1,2-Bis(triethoxysilyl)ethane, 95%

Product Code. 15457008
Change view
Click to view available options
Quantity:
5 mL
25 mL
Unit Size:
25mL
5mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15457008 25 mL 25mL
15447008 5 mL 5mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15457008 Supplier Thermo Scientific Alfa Aesar Supplier No. H60530.AC
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 16068-37-4 | C14H34O6Si2 | 354.59 g/mol

1,2-Bis(triethoxysilyl)ethane is used as coating auxiliary agents, leather auxiliary agents, plastic auxiliary agents, surfactants and textile auxiliary agents. It is used to prepare mesoporous organosilica materials.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1,2-Bis(triethoxysilyl)ethane is used as coating auxiliary agents, leather auxiliary agents, plastic auxiliary agents, surfactants and textile auxiliary agents. It is used to prepare mesoporous organosilica materials.

Solubility
Soluble in alcohol, aromatic and aliphatic hydrocarbons. Hydrolyzes with water.

Notes
Incompatible with oxidizing agents. Store in a cool, dry conditions in a well sealed container. Store at room temperature.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 16068-37-4
Molecular Formula C14H34O6Si2
Molecular Weight (g/mol) 354.59
MDL Number MFCD00239362
InChI Key IZRJPHXTEXTLHY-UHFFFAOYSA-N
Synonym 1,2-bis triethoxysilyl ethane, 4,4,7,7-tetraethoxy-3,8-dioxa-4,7-disiladecane, 3,8-dioxa-4,7-disiladecane, 4,4,7,7-tetraethoxy, ethylenebis triethoxysilane, bis triethoxysilyl ethane, hexaethoxydisilethylene, bis 1,2-triethoxysilyl ethane, 1,2-bis triethoxysilyl ethane,, triethoxysilyl ethyl triethoxysilane, triethoxy 2-triethoxysilylethyl silane
PubChem CID 85266
IUPAC Name 4,4,7,7-tetraethoxy-3,8-dioxa-4,7-disiladecane
SMILES CCO[Si](CC[Si](OCC)(OCC)OCC)(OCC)OCC

Specifications

Boiling Point 119°C
Quantity 25 mL
Linear Formula [-CH2Si(OC2H5)3]2
Flash Point 113°C (235°F)
Solubility Information Soluble in alcohol,aromatic and aliphatic hydrocarbons. Hydrolyzes with water.
Formula Weight 354.59
Percent Purity 95%
Refractive Index 1.411
Density 0.958
Chemical Name or Material 1,2-Bis(triethoxysilyl)ethane
compliance-icons compliance-icons
Product Identifier
  • 1,2-Bis(triethoxysilyl)ethane
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 3
  • Acute toxicity Category 4
  • LONG-TERM AQUATIC HAZARD Chronic 3
  • Specific target organ toxicity Category 1
Hazard Statement
  • H301-Toxic if swallowed.
  • H312-Harmful in contact with skin.
  • H372-Causes damage to organs.
  • H412-Harmful to aquatic life with long lasting effects.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
Supplemental information
  • MIXTURE LIST-Contains : 1,2-Bis(triethoxysilyl)ethane

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.