missing translation for 'onlineSavingsMsg'
Learn More

1,2-Bis(trimethylsiloxy)ethane, 98%

Product Code. 15450789
Change view
Click to view available options
Quantity:
10 g
50 g
Unit Size:
10g
50g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15450789 50 g 50g
15440789 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15450789 Supplier Thermo Scientific Alfa Aesar Supplier No. H63857.18
 Restricted Item
Estimated Shipment: 12-10-2026
Add to basket
This item is not returnable. View return policy

CAS: 7381-30-8 | C8H22O2Si2 | 206.43 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7381-30-8
Molecular Formula C8H22O2Si2
Molecular Weight (g/mol) 206.43
MDL Number MFCD00009640
InChI Key JGWFUSVYECJQDT-UHFFFAOYSA-N
Synonym 1,2-bis trimethylsiloxy ethane, 1,2-bis trimethylsilyloxy ethane, 2,2,7,7-tetramethyl-3,6-dioxa-2,7-disilaoctane, ethylene glycol bis trimethylsilyl ether, ethylenedioxybis trimethylsilane, 3,6-dioxa-2,7-disilaoctane, 2,2,7,7-tetramethyl, 1,2-bis-trimethylsilyloxy ethane, 1,2-bis-trimethylsilanyloxy-ethane, ethylene glycol, di-tms, acmc-1bh50
PubChem CID 81858
SMILES C[Si](C)(C)OCCO[Si](C)(C)C

Specifications

Boiling Point 165°C to 171°C
Quantity 50 g
Linear Formula (CH3)3SiOCH2CH2OSi(CH3)3
UN Number UN1993
Beilstein 1744217
Flash Point 51°C (124°F)
IUPAC Name trimethyl(2-trimethylsilyloxyethoxy)silane
Formula Weight 206.43
Percent Purity 98%
Refractive Index 1.403
Sensitivity Moisture sensitive
Density 0.842
Chemical Name or Material 1,2-Bis(trimethylsiloxy)ethane
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.