missing translation for 'onlineSavingsMsg'
Learn More

1-(2-Bromoethoxy)-2-bromobenzene, 95%, Thermo Scientific™

Product Code. 10095982 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass bottle
Unit Size:
25g
5g
Product Code. Quantity Packaging unitSize
10095982 5 g Glass bottle 5g
10177542 25 g Glass bottle 25g
2 options
This item is not returnable. View return policy

Product Code. 10095982

Brand: Thermo Scientific Acros 379220050

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Chemical Identifiers

CAS 18800-28-7
Molecular Formula C8H8Br2O
Molecular Weight (g/mol) 279.96
MDL Number MFCD00634301
InChI Key LOVVQTBEUIBASP-UHFFFAOYSA-N
Synonym 1-bromo-2-2-bromoethoxy benzene, 1-2-bromoethoxy-2-bromobenzene, o-bromo-2-bromoethoxybenzene, benzene, 1-bromo-2-2-bromoethoxy
PubChem CID 2734928
IUPAC Name 1-bromo-2-(2-bromoethoxy)benzene
SMILES BrCCOC1=CC=CC=C1Br

Specifications

CAS Min % 94.0
CAS Max % 100.0
Melting Point 35.0°C to 36.0°C
Color Colorless to Yellow
Boiling Point 160.0°C to 162.0°C (16.0 mmHg)
Infrared Spectrum Authentic
Assay Percent Range 94% min. (GC)
Packaging Glass bottle
Linear Formula BrC6H4O(CH2)2Br
Quantity 5 g
Formula Weight 279.96
Percent Purity 95%
Physical Form Liquid After Melting
Chemical Name or Material 1-(2-Bromoethoxy)-2-bromobenzene
Show More Show Less
compliance-icons
Product Identifier
  • 1-(2-Bromoethoxy)-2-bromobenzene
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.