missing translation for 'onlineSavingsMsg'
Learn More

1-(2-Bromoethoxy)-4-fluorobenzene, 97+%

Product Code. 15459117
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
Unit Size:
1g
250mg
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15459117 5 g 5g
15469117 250 mg 250mg
15449117 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15459117 Supplier Thermo Scientific Alfa Aesar Supplier No. H52403.06
 Restricted Item
Estimated Shipment: 15-09-2026
Add to basket
This item is not returnable. View return policy

CAS: 332-48-9 | C8H8BrFO | 219.05 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 332-48-9
Molecular Formula C8H8BrFO
Molecular Weight (g/mol) 219.05
MDL Number MFCD00044739
InChI Key JXSPKRUNMHMICQ-UHFFFAOYSA-N
Synonym 1-2-bromoethoxy-4-fluorobenzene, 4-fluorophenoxy-ethylbromide, 2-bromoethyl 4-fluorophenyl ether, 4-fluorophenoxyethylbromide, 4-2-bromoethoxy fluorobenzene, 4-fluorophenoxyethyl bromide, beta-bromo-4-fluorophenetole, 2-4-fluorophenoxy ethyl bromide, benzene, 1-2-bromoethoxy-4-fluoro, p-fluorophenoxy ethylbromide
PubChem CID 2064171
IUPAC Name 1-(2-bromoethoxy)-4-fluorobenzene
SMILES FC1=CC=C(OCCBr)C=C1

Specifications

Quantity 5 g
Formula Weight 219.05
Percent Purity ≥97%
Chemical Name or Material 1-(2-Bromoethoxy)-4-fluorobenzene

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.