missing translation for 'onlineSavingsMsg'
Learn More

1-(2-Bromoethyl)-4-nitrobenzene, 98%

Product Code. 11477617
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11477617 5 g 5g
11487617 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11477617 Supplier Thermo Scientific Alfa Aesar Supplier No. A15677.06
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 5339-26-4 | C8H8BrNO2 | 230.061 g/mol

1-(2-Bromoethyl)-4-nitrobenzene is an electrophile used as a substrate and activator for theta class gutathione transferase (GSTT-1). It is also used as a marker in for the θ-class of glutathione S-transferases (GSTs).

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1-(2-Bromoethyl)-4-nitrobenzene is an electrophile used as a substrate and activator for theta class gutathione transferase (GSTT-1). It is also used as a marker in for the θ-class of glutathione S-transferases (GSTs).

Solubility
Soluble in water (partly) and methanol.

Notes
Store in cool, well ventilated area. Keep container tightly closed. Store away form oxidizing agent.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 5339-26-4
Molecular Formula C8H8BrNO2
Molecular Weight (g/mol) 230.061
MDL Number MFCD00007386
InChI Key NTURQZFFJDCTMZ-UHFFFAOYSA-N
Synonym 4-nitrophenethyl bromide, 1-2-bromoethyl-4-nitrobenzene, benzene, 1-2-bromoethyl-4-nitro, 4-nitrophenethylbromide, p-nitrophenethyl bromide, 2-4-nitrophenyl ethyl bromide, 4-2-bromoethyl nitrobenzene, 4-nitrophenylethyl bromide, .beta.-p-nitrophenyl ethyl bromide, phenethylbromide4nitro
PubChem CID 79266
IUPAC Name 1-(2-bromoethyl)-4-nitrobenzene
SMILES C1=CC(=CC=C1CCBr)[N+](=O)[O-]

Specifications

Melting Point 67°C to 71°C
Boiling Point 116°C (0.2 mmHg)
Quantity 5 g
Beilstein 1909321
Solubility Information Soluble in water (partly) and methanol.
Formula Weight 230.07
Percent Purity 98%
Chemical Name or Material 1-(2-Bromoethyl)-4-nitrobenzene

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.